missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRSF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584300
This item is not returnable.
View return policy
Description
Immunogen sequence: KSRSVSRSPV PEKSQKRGSS SRSKSPA Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 96%.
SR proteins are a highly conserved family of arginine/serine-rich, spliceosome-associated phosphoproteins essential for metazoan pre-mRNA splicing. SR proteins act early in splicing by promoting splice site recognition and spliceosome assembly. SR proteins also play a regulatory role because they can determine alternative splice site usage in vivo and in vitro. SR proteins appear to be recruited from nuclear 'speckles', in which they are concentrated to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.
Specifications
| SRSF5 | |
| Polyclonal | |
| Unconjugated | |
| SRSF5 | |
| Cl-4; Delayed-early protein HRS; Hrs; Insulin-induced growth response protein CL-4; pre-mRNA-splicing factor SRP40; serine and arginine rich splicing factor 5; serine/arginine-rich splicing factor 5; SFRS5; splicing factor arginine/serine-rich 5 (SRp40 HRS); splicing factor, arginine/serine-rich 5; splicing factor, arginine/serine-rich 5 (SRp40, HRS); SR splicing factor 5; SRp40; Srsf5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6430 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13243 | |
| SRSF5 | |
| Recombinant protein corresponding to Human SRSF5. Recombinant protein control fragment (Product #RP-106710). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction