missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRSF5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15798211 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15798211 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15798211 Supplier Invitrogen™ Supplier No. PA560383

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KELCSERVTI EHARARSRGG RGRGRYSDRF SSRRPRNDRR NAPPVRTE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

SR proteins are a highly conserved family of arginine/serine-rich, spliceosome-associated phosphoproteins essential for metazoan pre-mRNA splicing. SR proteins act early in splicing by promoting splice site recognition and spliceosome assembly. SR proteins also play a regulatory role because they can determine alternative splice site usage in vivo and in vitro. SR proteins appear to be recruited from nuclear 'speckles', in which they are concentrated to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRSF5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRSF5
Gene Accession No. O35326, Q09167, Q13243
Gene Alias Cl-4; Delayed-early protein HRS; Hrs; Insulin-induced growth response protein CL-4; pre-mRNA-splicing factor SRP40; serine and arginine rich splicing factor 5; serine/arginine-rich splicing factor 5; SFRS5; splicing factor arginine/serine-rich 5 (SRp40 HRS); splicing factor, arginine/serine-rich 5; splicing factor, arginine/serine-rich 5 (SRp40, HRS); SR splicing factor 5; SRp40; Srsf5
Gene Symbols SRSF5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SRSF5. Recombinant protein control fragment (Product #RP-100370).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20384, 29667, 6430
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.