missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRSF1 Recombinant Rabbit Monoclonal Antibody (JB40-76)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534705
This item is not returnable.
View return policy
Description
Positive Control: Mouse heart tissue lysate, mouse liver tissue lysate, K562, A431, human lung cancer tissue, human tonsil tissue, human breast tissue, mouse colon tissue, rat epididymis tissue.
UBE2E2 catalyzes the covalent attachment of ubiquitin to other proteins.
Specifications
| SRSF1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q07955 | |
| SRSF1 | |
| Synthetic peptide within Human SF2 aa 1-50. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JB40-76 | |
| Unconjugated | |
| SRSF1 | |
| 1110054N12Rik; 5730507C05Rik; 6330415C05Rik; AI482334; Alternative-splicing factor 1; ASF; ASF/SF2; ASF-1; AW491331; OK/SW-cl.3; pre-mRNA-splicing factor SF2, P33 subunit; pre-mRNA-splicing factor SRp30a; serine and arginine rich splicing factor 1; serine/arginine-rich splicing factor 1; SF2; SF2p33; SFRS1; splicing factor 2; splicing factor, arginine/serine-rich 1; splicing factor, arginine/serine-rich 1 (ASF/SF2); splicing factor, arginine/serine-rich 1B; splicing factor, arginine/serine-rich, 30-KD, A; SR splicing factor 1; SRp30a; Srsf1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6426 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction