missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRSF1 Monoclonal Antibody (103)

Mouse Monoclonal Antibody
Brand: Invitrogen™ 324600
This item is not returnable.
View return policy
Description
SRSF1 Monoclonal Antibody for ICC/IF, Western Blot
UBE2E2 catalyzes the covalent attachment of ubiquitin to other proteins.
Specifications
| SRSF1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| Q07955, Q6PDM2 | |
| SRSF1 | |
| Recombinant SF2/ASF protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| 103 | |
| Unconjugated | |
| SRSF1 | |
| 1110054N12Rik; 5730507C05Rik; 6330415C05Rik; AI482334; Alternative-splicing factor 1; ASF; ASF/SF2; ASF-1; AW491331; OK/SW-cl.3; pre-mRNA-splicing factor SF2, P33 subunit; pre-mRNA-splicing factor SRp30a; serine and arginine rich splicing factor 1; serine/arginine-rich splicing factor 1; SF2; SF2p33; SFRS1; splicing factor 2; splicing factor, arginine/serine-rich 1; splicing factor, arginine/serine-rich 1 (ASF/SF2); splicing factor, arginine/serine-rich 1B; splicing factor, arginine/serine-rich, 30-KD, A; SR splicing factor 1; SRp30a; Srsf1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 110809, 6426, 689890 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction