missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRrp35 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | SRrp35 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SRrp35 Polyclonal specifically detects SRrp35 in Human samples. It is validated for Western Blot.Specifications
| SRrp35 | |
| Polyclonal | |
| Rabbit | |
| Q8WXF0 | |
| 135295 | |
| Synthetic peptides corresponding to SRRP35 The peptide sequence was selected from the middle region of SRRP35. Peptide sequence RSRSKSLQKRSKSIGKSQSSSPQKQTSSGTKSRSHGRHSDSIARSPCKSP. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 35 kDa SR repressor protein, FLJ14459, serine/arginine-rich splicing factor 12, serine-arginine repressor protein (35 kDa), SFRS13B, SFRS19FLJ41221, Splicing factor, arginine/serine-rich 13BFLJ33484, Splicing factor, arginine/serine-rich 19, SR splicing factor 12, SRRP35, SRrp35RP11-63L7.3 | |
| SRSF12 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title