missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRRM3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA549231

Product Code. 15663525

  • £439.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SRRM3 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SRRM3
Polyclonal
Unconjugated
SRRM3
2900083I11Rik; AB041661; MNCb-4760; RGD1307391; serine/arginine repetitive matrix 3; serine/arginine repetitive matrix protein 3; SRm300-like; Srrm2l; SRRM2-like protein; Srrm3
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
222183, 58212
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
2 mg/mL
PBS with 0.09% sodium azide; pH 7.4
A6NNA2, Q80WV7
SRRM3
KLH conjugated synthetic peptide between 264-291 amino acids from the Central region of human SRRM3.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.