missing translation for 'onlineSavingsMsg'
Learn More

SRRM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18381756 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381756 20 μL 20µL
18394976 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18381756 Supplier Novus Biologicals Supplier No. NBP30392320UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SRRM2 Polyclonal specifically detects SRRM2 in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRRM2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Cwc21, CWF21, DKFZp667L0421,300 kDa nuclear matrix antigen, DKFZp686O15166,300-KD, FLJ21926, FLJ22250, HSPC075, KIAA0324, MGC40295, RNA binding protein, ser/Arg-related nuclear matrix protein of 300 kDa, serine/arginine repetitive matrix 2, serine/arginine repetitive matrix protein 2, serine/arginine-rich splicing factor-related nuclear matrix protein of 300 kDa, splicing coactivator subunit SRm300, SRL300, SRm300, SR-related nuclear matrix protein of 300 kDa, taxREB803, tax-responsive enhancer element-binding protein 803
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SRRM2 (NP_057417.3).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23524
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.