Learn More
Invitrogen™ SRRM2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5118900
Product Code. 17750314
  • £439.00 / 100µL
Description
Antibody detects endogenous levels of total SRRM2.
SRRM2 is involved in pre-mRNA splicing. It may function at or prior to the first catalytic step of splicing at the catalytic center of the spliceosome and may do so by stabilizing the catalytic center or the position of the RNA substrate. It binds to RNA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SRRM2
Polyclonal
Unconjugated
SRRM2
300 kDa nuclear matrix antigen; 300-KD; 5033413A03Rik; AA410130; Cwc21; CWF21; HSPC075; Kiaa0324; mKIAA0324; RNA binding protein; ser/Arg-related nuclear matrix protein of 300 kDa; serine/arginine repetitive matrix 2; serine/arginine repetitive matrix protein 2; Serine/arginine-rich splicing factor-related nuclear matrix protein of 300 kDa; splicing coactivator subunit SRm300; SRL300; SRm300; SR-related nuclear matrix protein of 300 kDa; SRRM2; taxREB803; Tax-responsive enhancer element-binding protein 803; testicular secretory protein Li 53
Rabbit
Affinity chromatography
RUO
23524, 75956
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8BTI8, Q9UQ35
SRRM2
A synthesized peptide derived from human SRRM2(Accession Q9UQ35), corresponding to amino acid residues L2037-R2087.
100 μL
Primary
Human, Mouse