missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRRM2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-05971CL1
This item is not returnable.
View return policy
Description
SRRM2 Polyclonal antibody specifically detects SRRM2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence
Specifications
| SRRM2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 23524 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| Cwc21, CWF21, DKFZp667L0421,300 kDa nuclear matrix antigen, DKFZp686O15166,300-KD, FLJ21926, FLJ22250, HSPC075, KIAA0324, MGC40295, RNA binding protein, ser/Arg-related nuclear matrix protein of 300 kDa, serine/arginine repetitive matrix 2, serine/arginine repetitive matrix protein 2, serine/arginine-rich splicing factor-related nuclear matrix protein of 300 kDa, splicing coactivator subunit SRm300, SRL300, SRm300, SR-related nuclear matrix protein of 300 kDa, taxREB803, tax-responsive enhancer element-binding protein 803 | |
| Produced in rabbits immunized with E. coli-derived Human SRRM2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction