missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRPX2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£150.00
Specifications
| Antigen | SRPX2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
SRPX2 Polyclonal specifically detects SRPX2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SRPX2 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| NP_055282 | |
| 27286 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human SRPX2. The immunogen is located within the first 50 amino acids of SRPX2. Amino Acid Squence: SGYYPDESYNEVYAEE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| BPP, CBPS, RESDX, SRPULPMGX, sushi repeat-containing protein SRPX2, sushi-repeat containing protein, X-linked 2, sushi-repeat protein up-regulated in leukemia, sushi-repeat-containing protein, X-linked 2 | |
| SRPX2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title