missing translation for 'onlineSavingsMsg'
Learn More

SRPRB Antibody (OTI1D9), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18401367 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401367 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401367 Supplier Novus Biologicals Supplier No. NBP274350MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SRPRB Monoclonal antibody specifically detects SRPRB in Human, Mouse, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRPRB
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1D9
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias APMCF1, Protein APMCF1, signal recognition particle receptor subunit beta, signal recognition particle receptor, B subunit, signal recognition particle receptor, beta subunit, SR-beta
Host Species Mouse
Immunogen Full length human recombinant protein of human SRPRB(NP_067026) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58477
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.