missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
SRPRB Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP2-66584
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SRPRB Polyclonal specifically detects SRPRB in Canine, Chicken samples. It is validated for Western Blot, Immunohistochemistry.
Tekniske data
| SRPRB | |
| Polyclonal | |
| Western Blot 15 ug/ml, Immunohistochemistry 1:10 - 1:500 | |
| APMCF1, Protein APMCF1, signal recognition particle receptor subunit beta, signal recognition particle receptor, B subunit, signal recognition particle receptor, beta subunit, SR-beta | |
| Sheep | |
| Ammonium sulfate precipitation | |
| Primary | |
| Antibody will not detect SRPRB protein bound to SRPRA | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| PBS with 0.08% Sodium Azide | |
| SRPRB | |
| Synthetic peptide corresponding to amino acids 246-265 derived from the carboxyl terminal of the canine SRPRB subunit. | |
| 250 μg | |
| 58477 | |
| Canine, Chicken | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion