missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRPR Polyclonal Antibody
GREENER_CHOICE

Product Code. 13442407
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13442407 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13442407 Supplier Invitrogen™ Supplier No. PA528295

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, mouse liver. Predicted reactivity: Rat (99%), Chicken (92%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The gene encodes a subunit of the endoplasmic reticulum signal recognition particle receptor that, in conjunction with the signal recognition particle, is involved in the targeting and translocation of signal sequence tagged secretory and membrane proteins across the endoplasmic reticulum. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRPR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Srpra
Gene Accession No. P08240, Q9DBG7
Gene Alias 1300011P19Rik; D11Mgi27; Docking protein alpha; DP; DP-alpha; MGC17355; MGC3650; MGC9571; signal recognition particle receptor (docking protein); signal recognition particle receptor ('docking protein'); signal recognition particle receptor subunit alpha; Sralpha; SR-alpha; SRP receptor alpha subunit; SRP receptor subunit alpha; SRP-alpha; Srpr; SRPRA; Unknown (protein for MGC:128907)
Gene Symbols Srpr, Srpra
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SRPR. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6734, 67398
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.