missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRPR alpha Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93626-0.02ml
This item is not returnable.
View return policy
Description
SRPR alpha Polyclonal antibody specifically detects SRPR alpha in Human samples. It is validated for Western Blot
Specifications
| SRPR alpha | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Docking protein alpha, DP, DP-alpha, MGC17355, MGC3650, MGC9571, signal recognition particle receptor (docking protein), signal recognition particle receptor ('docking protein'), signal recognition particle receptor subunit alpha, Sralpha, SR-alpha, SRP-alpha | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human SRPR (NP_003130.2). MLDFFTIFSKGGLVLWCFQGVSDSCTGPVNALIRSVLLQERGGNNSFTHEALTLKYKLDNQFELVFVVGFQKILTLTYVDKLIDDVHRLFRDKYRTEIQQQSALSLLNGTFDFQNDFLRLLREAEESSKIRAPTTMKKFEDSEKAKKPVRSMIETRGEKPKEKAKNSKKKGAKKEGSDGPLATSKPVPAEKSGLPVGPENGVELSKEELIRRKREEFIQK | |
| 0.02 mL | |
| Signal Transduction | |
| 6734 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction