missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SRPK2, GST Tag Recombinant Protein

Product Code. 10432133
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
This item is not returnable. View return policy

Product Code. 10432133

Brand: Invitrogen™ PV4060

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

SRPK2, along with SRPK1, is most likely the cellular protein kinase mediating HBV core protein phosphorylation during viral infection. It therefore represents an important host cell target for therapeutic intervention in HBV infection. Promotes neuronal apoptosis by up-regulating cyclin-D1 (CCND1) expression. This is done by the phosphorylation of SRSF2, leading to the suppression of p53/TP53 phosphorylation thereby relieving the repressive effect of p53/TP53 on cyclin-D1 (CCND1) expression. Phosphorylates ACIN1, and redistributes it from the nuclear speckles to the nucleoplasm, resulting in cyclin A1 but not cyclin A2 up-regulation. Plays an essential role in spliceosomal B complex formation via the phosphorylation of DDX23/PRP28. Plays a negative role in the regulation of HBV replication through a mechanism not involving the phosphorylation of the core protein but by reducing the packaging efficiency of the pregenomic RNA (pgRNA) without affecting the formation of the viral core particles .
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78362
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 6733
Molecular Weight (g/mol) 105 kDa
Name Human SRPK2, GST Tag
Quantity 1 mg
Regulatory Status RUO - research use only
Gene Alias AW226533; AW492537; AW547358; H_RG152G17.1 A; H_RG152G17.1 b; mSRPK2; serine kinase SRPK2; serine/arginine-rich protein specific kinase 2; serine/arginine-rich protein-specific kinase 2; serine/arginine-rich splicing factor kinase 2; serine/threonine-protein kinase SRPK2; SFRS protein kinase 2; SFRSK2; SR protein kinase 2; SRPK2; SR-protein-specific kinase 2; SRSF protein kinase 2; SRSF protein kinase 2 C-terminal; SRSF protein kinase 2 N-terminal; Wbp6; WUGSC:H_RG152G17.1 A; WW domain binding protein 6
Common Name SRPK2
KinaseFamily SRPK Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.