missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRPK2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25417-1-AP-150UL
This item is not returnable.
View return policy
Description
EFHD2, also known as Swiprosin-1 or SWS1, is an EF-hand and coiled-coil-containing adaptor protein that plays a role in lymphocyte physiology. EFHD2 exhibits the highest expression in CD8+ T cells and immature B cells. It provides a membrane scaffold that is required for the Syk-, SLP-65-, and PLC-gamma2-dependent B-cell receptor (BCR)-induced calcium flux. EFHD2 may also regulate BCR-induced immature and primary B-cell apoptosis. It controls spontaneous apoptosis through the regulation of BCL2L1 abundance. Also, EFHD2 plays a role as negative regulator of the canonical NF-kappa-B-activating branch.Specifications
| SRPK2 | |
| Polyclonal | |
| Unconjugated | |
| SRPK2 | |
| SFRS protein kinase 2, SFRSK2, SR protein specific kinase 2, SRPK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6733 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P78362 | |
| SRPK2 | |
| SRPK2 Fusion Protein Ag17885 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction