Learn More
Invitrogen™ SRPK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596449
Description
Positive Samples: 293T, HepG2, MCF7, HeLa, THP-1, Rat testis, Mouse spleen; Cellular Location: Cytoplasm, Microsome, Microsome, Nucleus, Nucleus matrix Immunogen sequence: EEMEKESGPG QKRPNKQEES ESPVERPLKE NPPNKMTQEK LEESSTIGQD QTLMERDTEG GAAEINCNGV IEVINYTQNS NNETLRHKED LHNANDCDVQ NLNQESSFLS SQNGDSSTSQ ETDSCTPITS EVSDTMVCQS SSTVGQSFSE QHISQLQESI RAEIPCEDEQ EQEHNGPLDN KGKSTAGNFL V.
SRPK1 is a serine/threonine kinase that plays a role in the regulation of both constitutive and alternative splicing by regulating intracellular localization of splicing factors. SRPK1 localizes to the nucleus and the cytoplasm. It is thought to play a role in regulation of both constitutive and alternative splicing by regulating intracellular localization of splicing factors. A second alternatively spliced transcript variant for this gene has been described, but its full length nature has not been determined.
Specifications
| SRPK1 | |
| Polyclonal | |
| Unconjugated | |
| SRPK1 | |
| AU017960; OTTHUMP00000016277; serine/arginine-rich protein specific kinase 1; serine/arginine-rich protein-specific kinase 1; serine/arginine-rich splicing factor kinase 1; serine/threonine-protein kinase SRPK1; SFRS protein kinase 1; SFRSK1; SR protein-specific kinase 1; SRPK1; SR-protein-specific kinase 1; SRSF protein kinase 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20815, 361811, 6732 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.08 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O70551, Q96SB4 | |
| SRPK1 | |
| A synthetic peptide corresponding to a sequence within amino acids 290-480 of human SRPK1 (NP_0031283). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.