missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SRPK1 Antibody (2E8), Novus Biologicals™

Product Code. 18344549 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344549 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344549 Supplier Novus Biologicals™ Supplier No. H00006732M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SRPK1 Monoclonal antibody specifically detects SRPK1 in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRPK1
Applications ELISA, Western Blot, Sandwich ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 2E8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH38292
Gene Alias EC 2.7.11.1, Serine/arginine-rich protein-specific kinase 1, serine/arginine-rich splicing factor kinase 1, serine/threonine-protein kinase SRPK1, SFRS protein kinase 1SFRSK1, SR protein kinase 1, SR-protein-specific kinase 1, SRSF protein kinase 1
Host Species Mouse
Immunogen SRPK1 (AAH38292, 371 a.a. ∽ 470 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NETLRHKEDLHNANDCDVQNLNQESSFLSSQNGDSSTSQETDSCTPITSEVSDTMVCQSSSTVGQSFSEQHISQLQESIRAEIPCEDEQEQEHNGPLDNK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6732
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.