missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SRP9 Polyclonal antibody specifically detects SRP9 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SRP9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | ALURBP, MGC110981, signal recognition particle 9 kDa protein, signal recognition particle 9kD, signal recognition particle 9kDa |
| Host Species | Rabbit |
| Immunogen | A region between residue 36 to 86 of human Signal recognition particle 9 kDa protein. |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?