missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SRP68 Polyclonal antibody specifically detects SRP68 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SRP68 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | signal recognition particle 68 kDa protein, signal recognition particle 68kD, signal recognition particle 68kDa |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Signal Recognition Particle 68kDa using the numbering given in entry NP_055045.2 (GeneID 6730). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?