missing translation for 'onlineSavingsMsg'
Learn More

SRP68 Antibody (3A3), Novus Biologicals™

Product Code. 18362589 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362589 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362589 Supplier Novus Biologicals Supplier No. H00006730M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SRP68 Monoclonal antibody specifically detects SRP68 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRP68
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3A3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055045
Gene Alias signal recognition particle 68 kDa protein, signal recognition particle 68kD, signal recognition particle 68kDa
Host Species Mouse
Immunogen SRP68 (NP_055045.2, 531 a.a. ~ 627 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AILDANDAHQTETSSSQVKDNKPLVERFETFCLDPSLVTKQANLVHFPPGFQPIPCKPLFFDLALNHVAFPPLEDKLEQKTKSGLTGYIKGIFGFRS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6730
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.