missing translation for 'onlineSavingsMsg'
Learn More

SRp55 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18400906 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400906 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400906 Supplier Novus Biologicals Supplier No. NBP204142R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SRp55 Polyclonal antibody specifically detects SRp55 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRp55
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias B52, FLJ08061, pre-mRNA splicing factor SRP55, Pre-mRNA-splicing factor SRP55, serine/arginine-rich splicing factor 6, SFRS6, Splicing factor, arginine/serine-rich 6MGC5045, splicing factor, arginine/serine-rich, 55 kDa, SR splicing factor 6, SRP55arginine/serine-rich splicing factor 6
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to this antibody using the numbering given in entry NP_006266.2 (GeneID 6431).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6431
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.