missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRP19 Recombinant Rabbit Monoclonal Antibody (JE59-23)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544892
This item is not returnable.
View return policy
Description
SRP19 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
SRP19 belongs to the SRP19 family. It is signal-recognition-particle assembly and binds directly to 7S RNA and mediates binding of the 54 kDa subunit of the SRP.
Specifications
| SRP19 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P09132, Q9D7A6 | |
| SRP19 | |
| Recombinant protein within human SRP19 aa 1-100/144. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE59-23 | |
| Unconjugated | |
| SRP19 | |
| 2310020D23Rik; hypothetical protein LOC514960; signal recognition particle; signal recognition particle 19; signal recognition particle 19 kDa protein; signal recognition particle 19kD; signal recognition particle 19kDa; SRP19; zgc:63961 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 291685, 66384, 6728 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction