Learn More
SRP14 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11528-1-AP-20UL
 View more versions of this product
Product Code. 16898663
  • £135.00 / 20µL
Description
Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.
Specifications
Specifications
SRP14
Polyclonal
Unconjugated
SRP14
18 kDa Alu RNA binding protein, ALURBP, SRP14
Rabbit
Antigen Affinity Chromatography
RUO
296076, 6727
-20°C
Liquid
Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P37108
SRP14
SRP14 Fusion Protein Ag2099
20 μL
Primary
Human, Rat