missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRP14 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11528-1-AP-150UL
This item is not returnable.
View return policy
Description
Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.Specifications
| SRP14 | |
| Polyclonal | |
| Unconjugated | |
| SRP14 | |
| 18 kDa Alu RNA binding protein, ALURBP, SRP14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 296076, 6727 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P37108 | |
| SRP14 | |
| SRP14 Fusion Protein Ag2099 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction