missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRP14 Polyclonal Antibody

Description
Peptide sequence: KPIPKKGTVE GFEPADNKCL LRATDGKKKI STVVSSKEVN KFQMAYSNLL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 86%.
Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.
Specifications
Specifications
| Antigen | SRP14 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | Srp14 |
| Gene Accession No. | P37108 |
| Gene Alias | (homologous Alu RNA binding protein); 14kDa; 18 kDa Alu RNA-binding protein; ALURBP; AW536328; homologous Alu RNA binding protein; homologous Alu RNA-binding protein; hypothetical protein LOC619260; signal recognition particle 14; signal recognition particle 14 kDa protein; signal recognition particle 14kDa (homologous Alu RNA binding protein); SRP14; zgc:112290 |
| Gene Symbols | Srp14 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?