missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRP14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100113
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SRP14.
Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.
Specifications
| SRP14 | |
| Polyclonal | |
| Unconjugated | |
| Srp14 | |
| (homologous Alu RNA binding protein); 14kDa; 18 kDa Alu RNA-binding protein; ALURBP; AW536328; homologous Alu RNA binding protein; homologous Alu RNA-binding protein; hypothetical protein LOC619260; signal recognition particle 14; signal recognition particle 14 kDa protein; signal recognition particle 14kDa (homologous Alu RNA binding protein); SRP14; zgc:112290 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 296076, 6727 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P37108 | |
| Srp14 | |
| A synthesized peptide derived from human SRP14(Accession P37108), corresponding to amino acid residues A60-A110. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction