missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRMS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116368
This item is not returnable.
View return policy
Description
SRMS Polyclonal Antibody for Western Blot
SRMS (Srm) is a tyrosine protein kinase (SRC sub-family) that may be involved in the proliferation and differentiation of keratinocytes. Non-receptor tyrosine-protein kinase which phosphorylates DOK1 on tyrosine residues. Also phosphorylates KHDRBS1/SAM68 and VIM on tyrosine residues. Phosphorylation of KHDRBS1 is EGF-dependent.
Specifications
| SRMS | |
| Polyclonal | |
| Unconjugated | |
| SRMS | |
| A230069J08Rik; C20orf148; dJ697K14.1; PTK70; src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites; SRM; SRMS; Tyrosine-protein kinase Srms | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 20811 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q62270 | |
| SRMS | |
| KLH conjugated synthetic peptide between 113-147 amino acids from the Central region of Mouse Srms. | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction