missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576349
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The polyamines putrescine, spermine, and spermidine are ubiquitous polycationic mediators of cell growth and differentiation. SRM synthase is one of four enzymes in the polyamine-biosynthetic pathway and carries out the final step of spermidine biosynthesis. This enzyme catalyzes the conversion of putrescine to spermidine using decarboxylated S-adenosylmethionine as the cofactor.
Specifications
| SRM | |
| Polyclonal | |
| Unconjugated | |
| SRM | |
| AA407669; PAPT; Putrescine aminopropyltransferase; SpdST; SPDSY; Spermidine synthase; spermidine synthase-1; SPS1; Srm; SRML1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20810, 6723, 84596 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P19623, Q64674 | |
| SRM | |
| Recombinant protein of human SRM. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction