missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRM Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531341
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, Raji. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (88%), Xenopus laevis (93%), Rhesus Monkey (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The polyamines putrescine, spermine, and spermidine are ubiquitous polycationic mediators of cell growth and differentiation. SRM synthase is one of four enzymes in the polyamine-biosynthetic pathway and carries out the final step of spermidine biosynthesis. This enzyme catalyzes the conversion of putrescine to spermidine using decarboxylated S-adenosylmethionine as the cofactor.
Specifications
| SRM | |
| Polyclonal | |
| Unconjugated | |
| SRM | |
| AA407669; PAPT; Putrescine aminopropyltransferase; SpdST; SPDSY; Spermidine synthase; spermidine synthase-1; SPS1; Srm; SRML1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6723 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P19623 | |
| SRM | |
| Recombinant protein encompassing a sequence within the center region of human SRM. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction