missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SRM Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30219
This item is not returnable.
View return policy
Description
SRM Polyclonal antibody specifically detects SRM in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| SRM | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| EC 2.5.1, EC 2.5.1.16, Putrescine aminopropyltransferase, SPDSY, spermidine synthase, spermidine synthase-1, SPS1PAPT, SRML1 | |
| Between 100 and 150 | |
| 100 μg | |
| Protein Kinase | |
| 6723 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction