missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRI Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00310AF647
This item is not returnable.
View return policy
Description
SRI Polyclonal antibody specifically detects SRI in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| SRI | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| Alexa Fluor 647 | |
| calcium binding protein amplified in mutlidrug-resistant cells, CP22, CP-22, FLJ26259,22 kDa protein, H_RG167B05.1, SCN, sorcin, V19 | |
| Produced in rabbits immunized with purified, recombinant Human SRI (Accession#: P30626; Met1-Val198) | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 6717 | |
| Store at 4°C in the dark. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto