missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRGAP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545655
This item is not returnable.
View return policy
Description
Peptide sequence: SSVKKIEKMK EKRQAKYSEN KLKCTKARND YLLNLAATNA AISKYYIHDV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
GTPase-activating protein for RAC1 and perhaps Cdc42, but not for RhoA small GTPase. May attenuate RAC1 signaling in neurons. [UniProt].
Specifications
| SRGAP3 | |
| Polyclonal | |
| Unconjugated | |
| SRGAP3 | |
| AI452337; ARHGAP14; brain stress early protein Gbi; D130026O08Rik; Gbi; Kiaa0411; KIAA1156; MEGAP; mental disorder-associated GAP; mKIAA0411; RGD1563152; rho GTPase activating protein 14; rho GTPase-activating protein 14; SLIT-ROBO Rho GTPase activating protein 2; SLIT-ROBO Rho GTPase activating protein 3; SLIT-ROBO Rho GTPase-activating protein 3; SRGAP2; srGAP3; WAVE-associated Rac GTPase activating protein; WAVE-associated Rac GTPase-activating protein; WRP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9901 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O43295 | |
| SRGAP3 | |
| Synthetic peptide directed towards the N-terminal of human SRGAP3 (aa 213-262). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction