missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRGAP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | SRGAP2 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SRGAP2 Polyclonal specifically detects SRGAP2 in Human samples. It is validated for Western Blot.Specifications
| SRGAP2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FNBP2, Formin-binding protein 2, KIAA0456ARHGAP34formin binding protein 2, Rho GTPase-activating protein 34, SLIT-ROBO Rho GTPase activating protein 2, SLIT-ROBO Rho GTPase-activating protein 2, srGAP2, SRGAP3 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human SRGP2. Peptide sequence LKEAEKQEEKQIGKSVKQEDRQTPRSPDSTANVRIEEKHVRRSSVKKIEK | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 23380 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title