missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRGAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555792
This item is not returnable.
View return policy
Description
Immunogen sequence: WEGRHNGIDG LIPHQYIVVQ DTEDGVVERS SPKSEIEVIS EPPEEKVTAR AGASCPSGGH VADIYLANIN KQRKRPESGS IR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
This locus encodes a member of the SLIT-ROBO Rho GTPase activating protein family. The encoded protein stimulates GTPase activity of Rac1, and plays a role in cortical neuron development. This locus has several paralogs on human chromosome 1 resulting from segmental duplication. While this locus itself is conserved among various species, the paralogs are found only in the genus Homo, and not in the genomes of non-human great apes.
Specifications
| SRGAP2 | |
| Polyclonal | |
| Unconjugated | |
| SRGAP2 | |
| 9930124L22Rik; AI448945; ARHGAP34; FBP2; Fbp27; FBP-27; FNBP2; formin binding protein 2; formin-binding protein 2; Formin-binding protein 27; KIAA0456; RGD1566016; rho GTPase-activating protein 34; SLIT-ROBO GAP2; SLIT-ROBO Rho GTPase activating protein 2; SLIT-ROBO Rho GTPase-activating protein 2; Srgap2; SRGAP2A; srGAP3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23380 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75044 | |
| SRGAP2 | |
| Recombinant protein corresponding to Human SRGAP2. Recombinant protein control fragment (Product #RP-94433). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction