missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRGAP1, Mouse anti-Human, Clone: 5D2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00057522-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: PLDPETIAQDIEETMNTALNELRELERQSTAKHAPDVVLDTLEQVKNSPTPATSTESLSPLHNVALRSSEPQIRRSTSSSSDTMSTFKPMVAPRMGVQLSpecifications
| SRGAP1 | |
| Monoclonal | |
| Unconjugated | |
| SLIT-ROBO Rho GTPase activating protein 1 | |
| ARHGAP13/FLJ22166/KIAA1304 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 5D2 | |
| In 1x PBS, pH 7.4 | |
| NM_020762 | |
| SRGAP1 | |
| SRGAP1 (NP_065813, 952 a.a. ∼ 1050 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 57522 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction