missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRFBP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12792-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human SRFBP1.
SRFBP1 may be involved in regulating transcriptional activation of cardiac genes during the aging process. It may play a role in biosynthesis and/or processing of SLC2A4 in adipose cells.Specifications
| SRFBP1 | |
| Polyclonal | |
| Unconjugated | |
| SRFBP1 | |
| BUD22, P49, p49/STRAP, Rlb1, SRFBP1, STRAP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 153443, 67222 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8NEF9, Q9CZ91 | |
| Srfbp1 | |
| SRFBP1 Fusion Protein Ag3517 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction