missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRFBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560098
This item is not returnable.
View return policy
Description
Immunogen sequence: LKAAVQAFKE ARQNVAEVES SKNASEDNHS ENTLYSNDNG SNLQREATVI SEQKVKETKI LAKKPIHNSK EKIAKMEHGP KAVTIANSPS KP Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 51%.
SRFBP1 may be involved in regulating transcriptional activation of cardiac genes during the aging process. It may play a role in biosynthesis and/or processing of SLC2A4 in adipose cells.
Specifications
| SRFBP1 | |
| Polyclonal | |
| Unconjugated | |
| Srfbp1 | |
| 2810036K01Rik; BUD22; BUD22 homolog; FLJ25286; P49; p49/STRAP; RGD:1359398}; Rlb1; serum response factor binding protein 1; serum response factor-binding protein 1; SRFBP1; srfbp1 {ECO:0000312; SRF-dependent transcription regulation associated protein; SRF-dependent transcription regulation-associated protein; STRAP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 153443 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NEF9 | |
| Srfbp1 | |
| Recombinant protein corresponding to Human SRFBP1. Recombinant protein control fragment (Product #RP-96897). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction