missing translation for 'onlineSavingsMsg'
Learn More

SRFBP1 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18427833 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427833 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427833 Supplier Novus Biologicals Supplier No. NBP152655C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SRFBP1 Polyclonal antibody specifically detects SRFBP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRFBP1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias BUD22, BUD22 homolog, FLJ25286, p49, p49/STRAP, Rlb1, serum response factor binding protein 1, serum response factor-binding protein 1, SRF-dependent transcription regulation associated protein, SRF-dependent transcription regulation-associated protein, STRAP
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 379 and 429 of human SRF-Dependent Transcription Regulation-Associated Protein using the numbering given in entry NP_689759.2 (GeneID 153443).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 153443
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.