missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRF Mouse anti-Human, Clone: 1G8F4, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | SRF |
|---|---|
| Clone | 1G8F4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808905
|
Proteintech
66742-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898895
|
Proteintech
66742-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation.Specifications
| SRF | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| MCM1, Serum response factor, SRF | |
| SRF | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1G8F4 | |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P11831 | |
| 6722 | |
| SRF Fusion Protein Ag10386 | |
| Primary | |
| -20°C | |
| SRF |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title