missing translation for 'onlineSavingsMsg'
Learn More

SREBP2 Antibody (SREBP2/1579) - Azide and BSA Free, Novus Biologicals™

Product Code. 18390673 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390673 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390673 Supplier Novus Biologicals Supplier No. NBP308527

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SREBP2 Monoclonal specifically detects SREBP2 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SREBP2
Applications ELISA
Classification Monoclonal
Clone SREBP2/1579
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 10mM PBS
Gene Alias BHLHD2, bHLHd2sterol regulatory element-binding protein 2, Class D basic helix-loop-helix protein 2, SREBP2, SREBP2SREBP-2, sterol regulatory element binding transcription factor 2, Sterol regulatory element-binding transcription factor 2
Host Species Mouse
Immunogen Recombinant full-length human SREBP2 protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6721
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.