missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SREBP1 Monoclonal Antibody (2A4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516124
This item is not returnable.
View return policy
Description
In Western blot, a band can be seen at 125 kDa (precursor) and additional bands may be seen at 60-70 kDa (cleaved). Suggested positive control: IMR 5 and HeLa cells..
Sterol regulatory element-binding proteins (SREBPs) are transcription factors that are members of the basic helix-loop-helix leucine zipper family of DNA binding proteins. Three isoforms have been identified in mammalian tissues that vary in structure, regulation, and function. SREBP-1a and SREBP-1c (originally cloned as ADD1) are protein products of alternative promoter usage of the SREBP-1 gene. The third isoform is transcribed from a different gene, SREBP-2. SREBPs are present as 120 kDa inactive precursors in the endoplasmic reticulum (ER) membrane. Upon activation, the SREBP protein is translocated to the Golgi and proteolytic cleavage occurs resulting in a mature transcriptionally active 60-78 kDa fragment. In liver and adipose tissues, SREBPs have a significant influence on lipid and cholesterol accumulation by inducing the transcription of genes involved in these processes. While SREBP-1 is thought to be more important in regulating the expression of genes involved in triglyceride synthesis and accumulation, SREBP-2 has been more closely linked to those involved in cholesterol synthesis and accumulation.
Specifications
| SREBP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P36956, P56720, Q60416, Q9WTN3 | |
| SREBF1 | |
| 6 His-tag fusion protein of human SREBP1 corresponding to amino acids 301-407. | |
| 100 μL | |
| Primary | |
| Canine, Chicken, Hamster, Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Frozen), Western Blot | |
| 2A4 | |
| Unconjugated | |
| SREBF1 | |
| ADD1; ADD-1; Adipocyte determination- and differentiation-dependent factor 1; bHLHd1; Class D basic helix-loop-helix protein 1; D630008H06; I79_010155; nt-SREBP1; Processed sterol regulatory element-binding protein 1; SREBF1; SREBP1; SREBP-1; SREBP1a; SREBP-1a; SREBP1c; SREBP-1c; sterol regulatory element binding protein 1; sterol regulatory element binding protein-1; sterol regulatory element binding transcription factor 1; sterol regulatory element binding-protein 1; sterol regulatory element-binding protein 1; Sterol regulatory element-binding transcription factor 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100689018, 20787, 373915, 403475, 6720, 78968 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction