missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SREBP1 Antibody (SREBP1/1578), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08432CL1
This item is not returnable.
View return policy
Description
SREBP1 Monoclonal antibody specifically detects SREBP1 in Human samples. It is validated for ELISA
Specifications
| SREBP1 | |
| Monoclonal | |
| CoraFluor 1 | |
| BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1 | |
| Recombinant full-length human SREBP1 protein (Uniprot: P36956) | |
| 0.1 mL | |
| Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators | |
| 6720 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| ELISA | |
| SREBP1/1578 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction