missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SREBP1 Antibody (2A4), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-582V
This item is not returnable.
View return policy
Description
SREBP1 Monoclonal specifically detects SREBP1 in Human, Mouse, Rat, Canine, Chicken, Hamster, Monkey, Golden Syrian Hamster samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.
Specifications
| SREBP1 | |
| Monoclonal | |
| DyLight 405 | |
| P36956 | |
| SREBF1 | |
| 6 His-tag fusion protein of human SREBP1 corresponding to amino acids 301-407. [UniProt# P36956] | |
| 0.1 mL | |
| Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators | |
| 6720 | |
| Human, Mouse, Rat, Canine, Chicken, Hamster, Monkey, Golden Syrian Hamster | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoassay | |
| 2A4 | |
| Western Blot, Immunohistochemistry, Knockout Validated | |
| BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, mouse, rat and Golden Syrian Hamster. Canine reactivity reported in scientific literature (PMID: 23720350). Hamster reactivity reported in scientific literature (PMID: 24393244). | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction