Learn More
SREBF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14088-1-AP-150UL
 View more versions of this product
Product Code. 16842574
  • £349.00 / 150µL
Description
This antibody can recognize the 125 kDa precursor form and the 68 kDa mature form of human SREBF1.
Sterol regulatory element-binding proteins (SREBPs) are transcription factors that are members of the basic helix-loop-helix leucine zipper family of DNA binding proteins. Three isoforms have been identified in mammalian tissues that vary in structure, regulation, and function. SREBP-1a and SREBP-1c (originally cloned as ADD1) are protein products of alternative promoter usage of the SREBP-1 gene. The third isoform is transcribed from a different gene, SREBP-2. SREBPs are present as 120 kDa inactive precursors in the endoplasmic reticulum (ER) membrane. Upon activation, the SREBP protein is translocated to the Golgi and proteolytic cleavage occurs resulting in a mature transcriptionally active 60-78 kDa fragment. In liver and adipose tissues, SREBPs have a significant influence on lipid and cholesterol accumulation by inducing the transcription of genes involved in these processes. While SREBP-1 is thought to be more important in regulating the expression of genes involved in triglyceride synthesis and accumulation, SREBP-2 has been more closely linked to those involved in cholesterol synthesis and accumulation.
Specifications
Specifications
SREBF1
Polyclonal
Unconjugated
SREBF1
bHLHd1, SREBF1, SREBP 1, SREBP 1c, SREBP1
Rabbit
Antigen Affinity Chromatography
RUO
6720
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.54 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P36956
SREBF1
SREBF1 Fusion Protein Ag5219
150 μL