missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRD5A2 Rabbit anti-Human, Mouse, Rat, Clone: 8A4H5, Novus Biologicals™
Rabbit Monoclonal Antibody
£173.00 - £406.00
Specifications
| Antigen | SRD5A2 |
|---|---|
| Clone | 8A4H5 |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18325552
|
Bio-Techne
NBP3-15900-20UL |
20 μg |
£173.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18009025
|
Novus Biologicals
NBP3-15900-100UL |
100 μg |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SRD5A2 Monoclonal antibody specifically detects SRD5A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| SRD5A2 | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 3-oxo-5-alpha-steroid 4-dehydrogenase 2,5 alpha-SR2, EC 1.3.99.5, MGC138457, S5AR 2, SR type 2, Steroid 5-alpha-reductase 2,3-oxo-5 alpha-steroid 4-dehydrogenase 2, steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta4-dehydrogenase alpha 2), Type II 5-alpha reductase | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SRD5A2 (P31213). MQVQCQQSPVLAGSATLVALGALALYVAKPSGYGKHTESLKPAATRLPARAAWFLQELPSFAVPAGILARQPLSLFGPPGTVLLGLFCVHYFHRTFVYSL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 8A4H5 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6716 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title