missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRD5A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525465
This item is not returnable.
View return policy
Description
SRD5A2 Polyclonal Antibody for Western Blot
This gene encodes a microsomal protein expressed at high levels in androgen-sensitive tissues such as the prostate. The encoded protein is active at acidic pH and is sensitive to the 4-azasteroid inhibitor finasteride. Deficiencies in this gene can result in male pseudohermaphroditism, specifically pseudovaginal perineoscrotal hypospadias (PPSH).
Specifications
| SRD5A2 | |
| Polyclonal | |
| Unconjugated | |
| SRD5A2 | |
| 3-oxo-5-alpha-steroid 4-dehydrogenase 2; 5 alpha-SR2; 5ART2; MGC138457; S5AR 2; SR type 2; SRD5A2; ST5AR2; steroid 5 alpha reductase type2; steroid 5 alpha-reductase 2; steroid 5-alpha-reductase 2; steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2); type 2; 4-ene-3-oxosteroid 5-alpha oxidoreductase; type II 5-alpha reductase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 6716, 94224 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P31213, Q99N99 | |
| SRD5A2 | |
| KLH conjugated synthetic peptide between 153-182 amino acids from the central region of human SRD5A2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction