missing translation for 'onlineSavingsMsg'
Learn More

SRD5A1 Antibody, Novus Biologicals™

Product Code. 18343754 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343754 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343754 Supplier Novus Biologicals Supplier No. NBP314482

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SRD5A1 Polyclonal antibody specifically detects SRD5A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRD5A1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Simple Western 20 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 3-oxo-5-alpha-steroid 4-dehydrogenase 1, EC 1.3.99.5,3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 1, S5AR 1, SR type 1, Steroid 5-alpha-reductase 1,5-alpha reductase, steroid 5-alpha-reductase type I, steroid-5-alpha-reductase, alpha polypeptide 1 (3-oxo-5 alpha-steroid delta4-dehydrogenase alpha 1)
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6715
Target Species Human
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Product Type Antibody
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.