missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRD5A1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £366.00
Specifications
| Antigen | SRD5A1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637780
|
Novus Biologicals
NBP2-93860-0.02ml |
0.02 mL |
£150.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665290
|
Novus Biologicals
NBP2-93860-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SRD5A1 Polyclonal antibody specifically detects SRD5A1 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| SRD5A1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.3), 50% glycerol | |
| 6715 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| 3-oxo-5-alpha-steroid 4-dehydrogenase 1, EC 1.3.99.5,3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 1, S5AR 1, SR type 1, Steroid 5-alpha-reductase 1,5-alpha reductase, steroid 5-alpha-reductase type I, steroid-5-alpha-reductase, alpha polypeptide 1 (3-oxo-5 alpha-steroid delta4-dehydrogenase alpha 1) | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human SRD5A1 (NP_001038.1). LIYPFLMRGGKPMPLLACTMAIMFCTCNGYLQSRYLSHCAVYADDWVTDPRFLIGFGLWLTGMLINIHSDHILRNLRKPGDTGYKIPRGGLFEYVTAANYF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title