missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRCRB4D Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564037
This item is not returnable.
View return policy
Description
Immunogen sequence: PGEAHFGPGR GPILLDNVKC RGEESALLLC SHIRWDAHNC DHSEDASVLC QP Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.
Specifications
| SRCRB4D | |
| Polyclonal | |
| Unconjugated | |
| SSC4D | |
| Four scavenger receptor cysteine-rich domains-containing protein; S4D-SRCRB; S4-SRCRB; scavenger receptor cysteine rich domain containing, group B (4 domains); scavenger receptor cysteine rich family member with 4 domains; scavenger receptor cysteine rich family, 4 domains; scavenger receptor cysteine-rich domain-containing group B protein; scavenger receptor cysteine-rich protein SRCRB-S4D; SRCRB4D; SRCRB-S4D; SSC4D | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 136853 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WTU2 | |
| SSC4D | |
| Recombinant protein corresponding to Human SRCRB4D. Recombinant protein control fragment (Product #RP-103456). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction